5-Oxo-1-(4-sulfophenyl)-4,5-dihydro-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A299309-1G A299309-25G A299309-5G
77002-486EA 21 USD
77002-486 77002-488 77002-490
5-Oxo-1-(4-sulfophenyl)-4,5-dihydro-1H-pyrazole-3-carboxylic acid 95%
5-Oxo-1-(4-sulfophenyl)-4,5-dihydro-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₆S
MDL Number: MFCD00020755
CAS Number: 118-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR