1-(4-Nitrophenyl)piperidin-4-one 98%

Supplier: Ambeed
Warning

A299230-1G A299230-250MG A299230-25G A299230-5G
76988-744EA 50.93 USD
76988-744 76988-746 76988-748 76988-750
1-(4-Nitrophenyl)piperidin-4-one 98%
1-(4-Nitrophenyl)piperidin-4-one

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₃
MW: 220.23 g/mol
MDL Number: MFCD00231570
CAS Number: 23499-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR