tert-Butyl 3-iodo-7-methoxy-1H-indazole-1-carboxylate 95%

Supplier: Ambeed
Warning

A299080-100MG A299080-10G A299080-1G A299080-250MG A299080-5G
77014-904EA 20.16 USD
77014-904 77014-906 77014-908 77014-910 77014-912
tert-Butyl 3-iodo-7-methoxy-1H-indazole-1-carboxylate 95%
tert-Butyl 3-iodo-7-methoxy-1H-indazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₅IN₂O₃
MW: 374.17 g/mol
CAS Number: 639084-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR