6-Chloroisoquinolin-3(2H)-one 96%

Supplier: Ambeed
Warning

A298670-100MG A298670-1G A298670-250MG
77014-898EA 25 USD
77014-898 77014-900 77014-902
6-Chloroisoquinolin-3(2H)-one 96%
6-Chloroisoquinolin-3(2H)-one

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MDL Number: MFCD19704646
CAS Number: 51463-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR