Ethyl-6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydro-3-pyridinecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-oxo-1,4,5,6-tetrahydro-2-(trifluoromethyl)pyridine-3-carboxylate, Ethyl 6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydropyridine-3-carboxylate

A297684-100MG A297684-10G A297684-1G A297684-250MG A297684-5G A297684-25G
77002-454EA 5 USD
77002-454 77002-456 77002-458 77002-460 77002-462 77601-330
Ethyl-6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydro-3-pyridinecarboxylate 97%
Ethyl-6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydro-3-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₀F₃NO₃
MW: 237.178 g/mol
MDL Number: MFCD00975005
CAS Number: 194673-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR