1H-Pyrrolo[3,2-b]pyridine-6-carboxylic acid 96%

Supplier: Ambeed
Warning

A297650-100MG A297650-1G A297650-250MG A297650-5G
77002-446EA 51.99 USD
77002-446 77002-448 77002-450 77002-452
1H-Pyrrolo[3,2-b]pyridine-6-carboxylic acid 96%
1H-Pyrrolo[3,2-b]pyridine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD09763670
CAS Number: 112766-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR