2-Ethoxy-1H-indole 95%

Supplier: Ambeed
Warning

A297588-100MG A297588-1G A297588-250MG
77014-886EA 106 USD
77014-886 77014-888 77014-890
2-Ethoxy-1H-indole 95%
2-Ethoxy-1H-indole

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
MDL Number: MFCD13178666
CAS Number: 1009-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR