7-Benzyl-3-oxa-7-azabicyclo[3.3.1]nonan-9-one 95%

Supplier: Ambeed
Warning

A297241-100MG A297241-1G A297241-250MG
76988-724EA 8 USD
76988-724 76988-726 76988-728
7-Benzyl-3-oxa-7-azabicyclo[3.3.1]nonan-9-one 95%
7-Benzyl-3-oxa-7-azabicyclo[3.3.1]nonan-9-one

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₂
MDL Number: MFCD17926553
CAS Number: 77716-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR