(S)-2-Amino-3-chloropropanoic acid 95%
Supplier: Ambeed
|
Warning
|
A297107-100MG
A297107-1G
A297107-250MG
A297107-5G
76988-714EA
10.61
USD
76988-714
76988-716
76988-718
76988-720
(S)-2-Amino-3-chloropropanoic acid 95%
(S)-2-Amino-3-chloropropanoic acid
Precaución: for Research Use Only
Formula:
C₃H₆ClNO₂ MW: 123.53 g/mol |
MDL Number:
MFCD01632068 CAS Number: 39217-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...