(R)-2-Methoxypropanoic acid 95%

Supplier: Ambeed
Warning

A296675-100MG A296675-1G A296675-250MG A296675-5G
77014-876EA 12 USD
77014-876 77014-878 77014-880 77014-882
(R)-2-Methoxypropanoic acid 95%
(R)-2-Methoxypropanoic acid

Precaución: for Research Use Only

Formula: C₄H₈O₃
MDL Number: MFCD01632582
CAS Number: 23943-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR