4-[2-[(3-Ethyl-4-methyl-2-oxo-3-pyrrolin-1-yl)carboxamido]ethyl]benzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: Glimepiride sulphonamide

A296669-1G A296669-5G A296669-100G A296669-25G
77002-432EA 6.37 USD
77002-432 77002-434 77672-506 77672-508
4-[2-[(3-Ethyl-4-methyl-2-oxo-3-pyrrolin-1-yl)carboxamido]ethyl]benzenesulfonamide 98%
4-[2-[(3-Ethyl-4-methyl-2-oxo-3-pyrrolin-1-yl)carboxamido]ethyl]benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₆H₂₁N₃O₄S
MW: 351.42 g/mol
MDL Number: MFCD02955393
CAS Number: 119018-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR