4-Bromo-6-chloro-2-methylpyridazin-3(2H)-one 95%

Supplier: Ambeed
Warning

A296532-1G A296532-250MG
77014-872EA 73 USD
77014-872 77014-874
4-Bromo-6-chloro-2-methylpyridazin-3(2H)-one 95%
4-Bromo-6-chloro-2-methylpyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂O
MDL Number: MFCD21364605
CAS Number: 1178884-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR