Quinoxalin-5-ylmethanamine 97%

Supplier: Ambeed
Warning

A296514-100MG A296514-250MG
77014-868EA 219.61 USD
77014-868 77014-870
Quinoxalin-5-ylmethanamine 97%
Quinoxalin-5-ylmethanamine

Precaución: for Research Use Only

Formula: C₉H₉N₃
MW: 159.18 g/mol
MDL Number: MFCD16170468
CAS Number: 933694-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR