5,12-Dibromo-2,9-bis(2-ethylhexyl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone 95%

Supplier: Ambeed
Warning

A296291-1G A296291-250MG
77014-864EA 114 USD
77014-864 77014-866
5,12-Dibromo-2,9-bis(2-ethylhexyl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone 95%
5,12-Dibromo-2,9-bis(2-ethylhexyl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone

Precaución: for Research Use Only

Formula: C₄₀H₄₀Br₂N₂O₄
MDL Number: MFCD27923071
CAS Number: 851786-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR