2-Bromo-4,6-dimethoxypyridine 95%

Supplier: Ambeed
Warning

A295981-100MG A295981-1G A295981-250MG A295981-5G
77014-852EA 46.68 USD
77014-852 77014-854 77014-856 77683-883
2-Bromo-4,6-dimethoxypyridine 95%
2-Bromo-4,6-dimethoxypyridine

Precaución: For research use only.

Formula: C₇H₈BrNO₂
MW: 218.05 g/mol
MDL Number: MFCD03695815
CAS Number: 276251-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR