3-(Imidazo[1,2-a]pyrimidin-2-yl)propanoic acid 95%

Supplier: Ambeed
Warning

A295716-100MG A295716-1G A295716-250MG
77032-420EA 137 USD
77032-420 77032-422 77032-424
3-(Imidazo[1,2-a]pyrimidin-2-yl)propanoic acid 95%
3-(Imidazo[1,2-a]pyrimidin-2-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₉N₃O₂
MDL Number: MFCD11520330
CAS Number: 956101-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR