5-Bromo-2-methoxybenzonitrile 97%

Supplier: Ambeed
Warning

A295684-100G A295684-250MG A295684-25G A295684-5G A295684-1G A295684-10G
77032-412EA 223 USD
77032-412 77032-414 77032-416 77032-418 77601-322 77672-502
5-Bromo-2-methoxybenzonitrile 97%
5-Bromo-2-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MDL Number: MFCD00143429
CAS Number: 144649-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR