5-Iodo-6-methyl-1H-indazole 97%

Supplier: Ambeed
Warning

A295651-1G A295651-250MG
77014-848EA 172.93 USD
77014-848 77014-850
5-Iodo-6-methyl-1H-indazole 97%
5-Iodo-6-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇IN₂
MW: 258.05 g/mol
MDL Number: MFCD11878568
CAS Number: 1034154-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR