2-Chloro-4-iodopyridine-3-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-iodonicotinaldehyde

A295519-100G A295519-10G A295519-1G A295519-250MG A295519-25G A295519-5G
77002-402EA 753.24 USD
77002-402 77002-404 77002-406 77002-408 77002-410 77002-412
2-Chloro-4-iodopyridine-3-carboxaldehyde 98%
2-Chloro-4-iodopyridine-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₃ClINO
MW: 267.45 g/mol
MDL Number: MFCD03095294
CAS Number: 153034-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR