Palbociclib 99%

Supplier: Ambeed
Danger

Synonyms: 6-Acetyl-8-cyclopentyl-5-methyl-2-((5-(piperazin-1-yl)pyridin-2-yl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one

A295334-10MG A295334-1G A295334-25MG A295334-5G A295334-250MG A295334-100MG A295334-25G
77032-406EA 6.37 USD
77032-406 77032-408 77032-410 77550-796 77571-818 77601-316 77668-316
Palbociclib 99%
Palbociclib

Precaución: for Research Use Only

Formula: C₂₄H₂₉N₇O₂
MW: 447.53 g/mol
MDL Number: MFCD11840850
CAS Number: 571190-30-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR