Boc-L-β-homoalanine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-β-HoAla-OH, Boc-β-HomoAla-OH

A295251-1G A295251-25G A295251-5G A295251-100G
77002-394EA 12.74 USD
77002-394 77002-396 77002-398 77593-798
Boc-L-β-homoalanine 97%
Boc-L-beta-homoalanine

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
MDL Number: MFCD00270345
CAS Number: 158851-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR