1-(2-Methoxy-6-methylphenyl)ethanone 98%

Supplier: Ambeed
Warning

A294683-1G A294683-5G
77014-836EA 455.13 USD
77014-836 77014-838
1-(2-Methoxy-6-methylphenyl)ethanone 98%
1-(2-Methoxy-6-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
MDL Number: MFCD22068670
CAS Number: 6161-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR