2-Phenyloxetane 95+%

Supplier: Ambeed
Warning

A294621-100MG A294621-1G A294621-250MG
76988-690EA 63 USD
76988-690 76988-692 76988-694
2-Phenyloxetane 95+%
2-Phenyloxetane

Precaución: for Research Use Only

Formula: C₉H₁₀O
MW: 134.17 g/mol
MDL Number: MFCD18909301
CAS Number: 4436-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR