(S)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid hydrochloride 98%

Supplier: Ambeed
Danger

A294611-100G A294611-10G A294611-1G A294611-250MG A294611-25G A294611-5G
77032-390EA 89.12 USD
77032-390 77032-392 77032-394 77032-396 77032-398 77032-400
(S)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid hydrochloride 98%
(S)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₂₁ClFN₃O₄
MW: 397.82 g/mol
MDL Number: MFCD03265511
CAS Number: 177325-13-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR