Methyl-4-bromothieno[2,3-c]pyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromothieno[2,3-c]pyridine-2-carboxylate

A294322-250MG A294322-5G
77032-372EA 29.71 USD
77032-372 77601-312
Methyl-4-bromothieno[2,3-c]pyridine-2-carboxylate 97%
Methyl-4-bromothieno[2,3-c]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂S
MW: 272.13 g/mol
MDL Number: MFCD08275124
CAS Number: 145325-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR