4-Chloro-6-(pyrrolidin-1-yl)pyrimidin-5-amine 95%

Supplier: Ambeed
Danger

A294263-100MG A294263-1G A294263-250MG
77002-374EA 43 USD
77002-374 77002-376 77002-378
4-Chloro-6-(pyrrolidin-1-yl)pyrimidin-5-amine 95%
4-Chloro-6-(pyrrolidin-1-yl)pyrimidin-5-amine

Precaución: for Research Use Only

Formula: C₈H₁₁ClN₄
MDL Number: MFCD04035510
CAS Number: 626217-76-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR