1-Methyltetrahydropyrimidin-2(1H)-one 95%

Supplier: Ambeed
Warning

A294181-1G A294181-250MG A294181-5G
77002-368EA 47 USD
77002-368 77002-370 77002-372
1-Methyltetrahydropyrimidin-2(1H)-one 95%
1-Methyltetrahydropyrimidin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O
MDL Number: MFCD00196814
CAS Number: 10166-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR