Anacetrapib ≥98%

Supplier: Ambeed
Warning

Synonyms: (4S,5R)-5-(3,5-Bis(trifluoromethyl)phenyl)-3-((4'-fluoro-5'-isopropyl-2'-methoxy-4-(trifluoromethyl)-[1,1'-biphenyl]-2-yl)methyl)-4-methyloxazolidin-2-one

A293882-10MG A293882-1MG A293882-25MG A293882-2MG A293882-50MG A293882-5MG
77032-360EA 10.61 USD
77032-360 77032-362 77032-364 77032-366 77032-368 77032-370
Anacetrapib ≥98%
Anacetrapib

Precaución: for Research Use Only

Formula: C₃₀H₂₅F₁₀NO₃
MW: 637.51 g/mol
MDL Number: MFCD16294903
CAS Number: 875446-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR