2,7-Diazaspiro[3.5]nonan-1-one 97%

Supplier: Ambeed
Warning

A293821-100MG A293821-250MG
77014-826EA 92 USD
77014-826 77014-828
2,7-Diazaspiro[3.5]nonan-1-one 97%
2,7-Diazaspiro[3.5]nonan-1-one

Precaución: for Research Use Only

Formula: C₇H₁₂N₂O
MDL Number: MFCD13180578
CAS Number: 1147422-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR