Methyl-2-iodo-5-methylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-Iodo-5-methylbenzoate

A293688-1G A293688-5G
77002-364EA 21.22 USD
77002-364 77002-366
Methyl-2-iodo-5-methylbenzoate 98%
Methyl 2-iodo-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉IO₂
MW: 276.07 g/mol
MDL Number: MFCD12198129
CAS Number: 103440-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR