tert-Butyl (2-amino-4-(thiophen-2-yl)phenyl)carbamate 95%

Supplier: Ambeed
Warning

A293536-1G A293536-250MG
76988-684EA 183.54 USD
76988-684 76988-686
tert-Butyl (2-amino-4-(thiophen-2-yl)phenyl)carbamate 95%
tert-Butyl (2-amino-4-(thiophen-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂O₂S
MW: 290.38 g/mol
MDL Number: MFCD24038830
CAS Number: 335255-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR