[4-(1-Methyl-1-phenylethyl)phenoxy]acetic acid 95%

Supplier: Ambeed
Warning

A293432-5G A293432-250MG
77032-358EA 330 USD
77032-358 77872-378
[4-(1-Methyl-1-phenylethyl)phenoxy]acetic acid 95%
[4-(1-Methyl-1-phenylethyl)phenoxy]acetic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₃
MDL Number: MFCD00423397
CAS Number: 70757-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR