4-Amino-6-methyl-4,5-dihydro-1,2,4-triazin-3(2H)-one hydrochloride 95%

Supplier: Ambeed
Warning

A292812-100MG A292812-1G A292812-250MG
77032-348EA 129 USD
77032-348 77032-350 77032-352
4-Amino-6-methyl-4,5-dihydro-1,2,4-triazin-3(2H)-one hydrochloride 95%
4-Amino-6-methyl-4,5-dihydro-1,2,4-triazin-3(2H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉ClN₄O
MDL Number: MFCD18072820
CAS Number: 158329-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR