3-(Benzyloxy)benzoic acid 96%

Supplier: Ambeed
Warning

Synonyms: 3-Benzyloxybenzoic acid

A292611-1G A292611-25G A292611-5G
77032-342EA 21.22 USD
77032-342 77032-344 77032-346
3-(Benzyloxy)benzoic acid 96%
3-(Benzyloxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
MDL Number: MFCD00527901
CAS Number: 69026-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR