3-Bromo-5-chloropyridine-2-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-chloropicolinonitrile

A292609-10G A292609-1G A292609-250MG A292609-25G A292609-5G
77032-332EA 199.45 USD
77032-332 77032-334 77032-336 77032-338 77032-340
3-Bromo-5-chloropyridine-2-carbonitrile 98%
3-Bromo-5-chloropyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₂BrClN₂
MW: 217.46 g/mol
MDL Number: MFCD12827549
CAS Number: 760207-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR