Uracil-5-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde, 5-Formyluracil

A292599-1G A292599-5G
77014-822EA 30.77 USD
77014-822 77014-824
Uracil-5-carboxaldehyde 95%
Uracil-5-carboxaldehyde

Precaución: for Research Use Only

Formula: C₅H₄N₂O₃
MW: 140.10 g/mol
MDL Number: MFCD00192185
CAS Number: 1195-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR