4,6-Dichloro-2-methylnicotinic acid 97%

Supplier: Ambeed
Warning

A292256-100MG A292256-250MG A292256-1G
77032-322EA 43 USD
77032-322 77032-324 77881-805
4,6-Dichloro-2-methylnicotinic acid 97%
4,6-Dichloro-2-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MDL Number: MFCD13191581
CAS Number: 693286-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR