(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Tyr(tBu)-OH, N-Boc-O-tert-butyl-L-tyrosine

A292086-100G A292086-10G A292086-1G A292086-25G A292086-5G A292086-500G
77032-312EA 118 USD
77032-312 77032-314 77032-316 77032-318 77032-320 77571-802
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 97%
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₇NO₅
MW: 337.4 g/mol
MDL Number: MFCD00065598
CAS Number: 47375-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR