(6-(Ethylsulfonyl)pyridin-3-yl)boronic acid 98%

Supplier: Ambeed
Warning

A291984-100MG A291984-1G A291984-250MG
77014-810EA 47 USD
77014-810 77014-812 77014-814
(6-(Ethylsulfonyl)pyridin-3-yl)boronic acid 98%
(6-(Ethylsulfonyl)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₄S
MDL Number: MFCD17015735
CAS Number: 1256345-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR