3-Chromanecarboxylic acid 96%

Supplier: Ambeed
Warning

Synonyms: Chroman-3-carboxylic acid

A291525-100MG A291525-1G A291525-1MG A291525-250MG A291525-25G A291525-5G
77014-794EA 18.04 USD
77014-794 77014-796 77014-798 77014-800 77014-802 77612-676
3-Chromanecarboxylic acid 96%
3-Chromanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
MDL Number: MFCD03783554
CAS Number: 115822-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR