Di(pyridin-3-yl)methanol 95%

Supplier: Ambeed
Warning

A291171-100MG A291171-1G A291171-250MG
76988-664EA 45 USD
76988-664 76988-666 76988-668
Di(pyridin-3-yl)methanol 95%
Di(pyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O
MDL Number: MFCD12808134
CAS Number: 89667-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR