Methyl 3-bromo-4-fluoro-1H-indazole-6-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A290673-100MG
A290673-250MG
77014-774EA
129.43
USD
77014-774
77014-776
Methyl 3-bromo-4-fluoro-1H-indazole-6-carboxylate 95%
Methyl 3-bromo-4-fluoro-1H-indazole-6-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₆BrFN₂O₂ MW: 273.06 g/mol |
MDL Number:
MFCD08273743 CAS Number: 885521-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...