(S)-5-Oxo-1-((S)-1-phenylethyl)pyrrolidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A290591-1G A290591-250MG A290591-5G
76988-652EA 73 USD
76988-652 76988-654 76988-656
(S)-5-Oxo-1-((S)-1-phenylethyl)pyrrolidine-3-carboxylic acid 97%
(S)-5-Oxo-1-((S)-1-phenylethyl)pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃
MDL Number: MFCD03427553
CAS Number: 173340-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR