5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(2-((trimethylsilyl)oxy)propan-2-yl)pyridine 98%

Supplier: Ambeed
Warning

A290314-250MG A290314-5G A290314-1G
76988-650EA 19 USD
76988-650 77571-794 77593-778
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(2-((trimethylsilyl)oxy)propan-2-yl)pyridine 98%

Precaución: for Research Use Only

Formula: C₁₇H₃₀BNO₃Si
MDL Number: MFCD12198135
CAS Number: 1228014-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR