3-Acetylphenanthrene 90%

Supplier: Ambeed
Warning

Synonyms: 1-(phenanthren-3-yl)ethanone, 1-(phenanthren-3-yl)ethan-1-one, Ethanone, 1-(3-phenanthrenyl)-

A290274-250MG A290274-25G
77032-280EA 25.47 USD
77032-280 77032-282
3-Acetylphenanthrene 90%
3-Acetylphenanthrene

Precaución: for Research Use Only

Formula: C₁₆H₁₂O
MW: 220.27 g/mol
MDL Number: MFCD00001169
CAS Number: 2039-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR