S-tert-Butyl-L-cysteine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Cys(tBu)-OH.HCl, H-Cys(tBu)-OH·HCl

A290228-100G A290228-1G A290228-250MG A290228-25G A290228-5G A290228-10G
77002-346EA 248 USD
77002-346 77002-348 77002-350 77002-352 77002-354 77601-290
S-tert-Butyl-L-cysteine hydrochloride 98%
S-tert-Butyl-L-cysteine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClNO₂S
MW: 213.72 g/mol
MDL Number: MFCD00037426
CAS Number: 2481-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR