3-Amino-4-bromo-5-methylpyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-5-methyl-1H-pyrazol-3-amine

A290117-1G A290117-250MG A290117-25G A290117-5G
77014-760EA 41.38 USD
77014-760 77014-762 77014-764 77014-766
3-Amino-4-bromo-5-methylpyrazole 98%
3-Amino-4-bromo-5-methylpyrazole

Precaución: for Research Use Only

Formula: C₄H₆BrN₃
MW: 176.02 g/mol
MDL Number: MFCD03412169
CAS Number: 1780-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR