1-(2,3-Dimethoxyphenyl)-N-methylmethanamine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A289949-250MG
A289949-5G
76988-642EA
31.83
USD
76988-642
77683-868
1-(2,3-Dimethoxyphenyl)-N-methylmethanamine hydrochloride 97%
1-(2,3-Dimethoxyphenyl)-N-methylmethanamine hydrochloride
Precaución: For research use only.
Formula:
C₁₀H₁₆ClNO₂ MW: 217.69 g/mol |
MDL Number:
MFCD07108392 CAS Number: 1158741-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...