5-Chloropyrimidine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A289876-100MG A289876-250MG A289876-1G
77014-754EA 20 USD
77014-754 77014-756 77892-577
5-Chloropyrimidine-4-carboxylic acid 95%
5-Chloropyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₂
MDL Number: MFCD09754103
CAS Number: 64224-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR