4-Bromo-N-methylbutan-1-amine hydrobromide 95%

Supplier: Ambeed
Warning

A289746-100MG A289746-250MG
76988-638EA 111 USD
76988-638 76988-640
4-Bromo-N-methylbutan-1-amine hydrobromide 95%
4-Bromo-N-methylbutan-1-amine hydrobromide

Precaución: for Research Use Only

Formula: C₅H₁₃Br₂N
MDL Number: MFCD27920606
CAS Number: 365432-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR